Skip to Content
Merck
All Photos(1)

Key Documents

375095

Millipore

Heparin, Sodium Salt, Porcine Intestinal Mucosa

Synonym(s):

Heparin, Sodium Salt, Porcine Intestinal Mucosa, HEP, Na

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.79

form

lyophilized

Quality Level

specific activity

≥160 USP units/mg dry wt

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤0.5 EU/mg Endotoxin

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

15-25°C

General description

M.W. 13,500-15,000. Note: 1 KU = 1000 units.
MW: 15,000 ± 2,000
Preservative-free, non-sterile. Highly sulfated dextrorotatory mucopolysaccharide with specific anticoagulant properties. Inhibits IP3-activated Ca2+ channel release of the endoplasmic reticulum. Reported to activate the ryanodine receptor. Note: 1 KU = 1000 units. MW: 15,000 ± 2,000.
Preservative-free, non-sterile. Highly sulfated dextrorotatory mucopolysaccharide with specific anticoagulant properties. Inhibits the IP3-activated Ca2+ channel release of the endoplasmic reticulum. Reported to activate the ryanodine receptor. Note: 1 KU = 1000 units.

Warning

Toxicity: Harmful (C)

Other Notes

Seuwen, K., and Boddeke, H.G. 1995. Br. J. Pharmacol.114, 1613.
Ehrlich, B.E., et al. 1994. Trends Pharmacol. Sci.15,145.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enrique Gómez et al.
Journal of proteome research, 19(3), 1169-1182 (2020-01-25)
Metabolomics may identify biomarkers in blood that differentiate pregnant from open embryo recipients. Fresh and vitrified/warmed, in vitro-produced embryos were transferred to Holstein recipients (discovery group). Recipient blood plasma collected on Day-0 (estrus) and Day-7 (before embryo transfer) were analyzed
Junxin Wang et al.
Bioconjugate chemistry, 29(11), 3768-3775 (2018-10-04)
We recently reported a real-time method to measure heparin in human whole blood based on the photoacoustic change of methylene blue (MB). Intriguingly, the MB behaved unlike other "turn on" photoacoustic probes-the absorbance decreased as the photoacoustic signal increased. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service