Skip to Content
Merck
All Photos(2)

Key Documents

1.46028

Millipore

Sabouraud Dextrose Agar

Heimplate, EP, JP, USP, Settle plate for long incubation, plate diam. 90 mm, sterile, single packed of 20 or 120 plates, suitable for air monitoring

Synonym(s):

SDA Plate, Settle plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

Agency

EP
JP
USP

Quality Level

sterility

sterile

form

plate (ready-to-use)

feature

ready-to-use
Settle plate for long incubation

manufacturer/tradename

Heimplate

packaging

single packed of 20 or 120 plates

technique(s)

microbiological culture: suitable

plate diam.

90 mm

pH

5.6 ( in H2O)

application(s)

air monitoring
microbiology
pharmaceutical

storage temp.

15-25°C

suitability

molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

Sabouraud Dextrose Agar in 90 mm settle plate with 30 mL filling volume for long incubation is designed for the detection of yeasts and molds in the air via active or passive air monitoring as well as personnel monitoring on fingerprints and gloves in controlled environments in the presence of disinfectant residues and also to check the cleanliness.
Sabouraud Dextrose Medium contains digests of animal tissues (peptones), which provide a nutritional source of amino acids and nitrogenous compounds for the growth of fungi and yeasts. Dextrose is added as the energy and carbon source. The low pH value of 5.4-5.8 favors fungal growth and inhibits contaminating bacteria from samples.

Application

Sabouraud Dextrose Agar settle plate is designed for the detection of the total count of yeasts and molds in air and from personnel in less critical environments.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service