Skip to Content
Merck
All Photos(1)

Key Documents

12-309

Sigma-Aldrich

HeLa Nuclear Extract

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105330
eCl@ss:
32011807
NACRES:
NA.41

biological source

human

Quality Level

form

liquid

manufacturer/tradename

Upstate®

concentration

2.0 mg/mL

technique(s)

western blot: suitable

shipped in

dry ice

General description

Nuclear extract prepared from human HeLa cells using a modified protocol of Dignam, et al., which contains a variety of DNA binding proteins and transcription factors.

Cell Line Description

HeLa

Quality

Routinely evaluated as a positive control for many of Upstates antibodies.

Physical form

RIPA sample buffer containing inhibitors. Frozen solution.

Storage and Stability

Stable for 6 months at -70°C from date of shipment.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J D Dignam et al.
Nucleic acids research, 11(5), 1475-1489 (1983-03-11)
We have developed a procedure for preparing extracts from nuclei of human tissue culture cells that directs accurate transcription initiation in vitro from class II promoters. Conditions of extraction and assay have been optimized for maximum activity using the major
Tabetha Sundin et al.
Biochimie, 94(12), 2639-2648 (2012-08-21)
Isoprenoids are recognized for their ability to suppress carcinogenic processes in vivo and in vitro. We previously established that the isoprenoid, perillyl alcohol, acted mechanistically on translation of specific proteins through modulation of mechanistic target of rapamycin (mTOR) signaling. Telomerase-the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service