Skip to Content
Merck
All Photos(1)

Key Documents

06-983

Sigma-Aldrich

Anti-GADS Antibody

Upstate®, from rabbit

Synonym(s):

Anti-GADS, Anti-GRAP-2, Anti-GRB2L, Anti-GRBLG, Anti-GRID, Anti-GRPL, Anti-GrbX, Anti-Grf40, Anti-Mona, Anti-P38

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GRAP2(9402)

Specificity

GADS

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 174-188 of murine GADS (GEEIRPSVNRKLSDH)

Application

Anti-GADS Antibody is an antibody against GADS for use in IP & WB.

Quality

routinely evaluated by immunoblot on Jurkat RIPA cell lysate

Target description

40kDa

Physical form

Format: Purified

Analysis Note

Control
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hematopoietic progenitor kinase 1 negatively regulates T cell receptor signaling and T cell-mediated immune responses.
Jr-Wen Shui, Jonathan S Boomer, Jin Han, Jun Xu, Gregory A Dement, Guisheng Zhou, Tse-Hua Tan
Nature Immunology null
Eri Ishikawa et al.
Nature communications, 7, 12756-12756 (2016-09-28)
Thymic selection shapes an appropriate T cell antigen receptor (TCR) repertoire during T cell development. Here, we show that a serine/threonine kinase, protein kinase D (PKD), is crucial for thymocyte positive selection. In T cell-specific PKD-deficient (PKD2/PKD3 double-deficient) mice, the
Quantitative proteomics analysis of signalosome dynamics in primary T cells identifies the surface receptor CD6 as a Lat adaptor-independent TCR signaling hub.
Roncagalli, R; Hauri, S; Fiore, F; Liang, Y; Chen, Z; Sansoni, A; Kanduri, K; Joly et al.
Nature Immunology null
Luminal leptin inhibits L-glutamine transport in rat small intestine: involvement of ASCT2 and B0AT1.
Ducroc R, Sakar Y, Fanjul C, Barber A, Bado A, Lostao MP
American Journal of Physiology: Gastrointestinal and Liver Physiology null
Juris A Grasis et al.
Molecular and cellular biology, 30(14), 3596-3609 (2010-05-12)
In vitro data have suggested that activation of the inducible T-cell kinase (ITK) requires an interaction with the adaptor protein SLP-76. One means for this interaction involves binding of the ITK SH3 domain to the polyproline-rich (PR) region of SLP-76.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service