Skip to Content
Merck
All Photos(1)

Key Documents

06-1005

Sigma-Aldrich

Anti-TRIC-A Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

transmembrane protein 38A, trimeric intracellular cation channel type A, Tmem38a, NET1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TMEM38A(79041)

General description

TRIC-A (trimeric intracellular cation channel type A), also known as Tmem38A, and sometimes NET1, is one of a number of TRIC channel subtypes that are believed to function as a Ca2+ counter-ion channel. TRIC-A is highly expressed in excitable cells and seems to work in coordination with Ca2+ release to balance transient negative potential. TRIC-A is located on the sarcoplasmic reticulum (SR) and is thought to be regulated by trans-membrane voltage. TRIC-A deficiency may lead to an abundance of Ca2+ and cause volatility in Ca2+ movement across the SR membrane.

Specificity

This antibody recognizes the cytoplasmic domain of TRIC-A.

Immunogen

Epitope: Cytoplasmic domain
KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human TRIC-A.

Application

Anti-TRIC-A Antibody detects level of TRIC-A & has been published & validated for use in WB, IH & IC.
Immunohistochemistry Analysis: A representative lot was used by an independent laboratory in IH. (Wilkie, G.S., et al. (2011). Mol Cell Proteomics. 10(1):M110.003129-1).

Immunocytochemistry Analysis: A representative lot was used by an independent laboratory in IC. (Wilkie, G.S., et al. (2011). Mol Cell Proteomics. 10(1):M110.003129-1).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
GPCR, cAMP/cGMP & Calcium Signaling

Quality

Evaluated by Western Blot in human fetal skeletal muscle tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected TRIC-A on 10 µg of human fetal skeletal muscle tissue lysate.

Target description

~33 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in PBS containing 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human fetal skeletal muscle tissue lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne T Bertrand et al.
Cells, 9(4) (2020-04-05)
LMNA encodes for Lamin A/C, type V intermediate filaments that polymerize under the inner nuclear membrane to form the nuclear lamina. A small fraction of Lamin A/C, less polymerized, is also found in the nucleoplasm. Lamin A/C functions include roles
Stefan Hintze et al.
Frontiers in physiology, 9, 1532-1532 (2018-11-15)
Myotonic dystrophy type 1 (DM1) is a multisystemic disorder with predominant myotonia and muscular dystrophy which is caused by CTG-repeat expansions in the DMPK gene. These repeat expansions are transcribed and the resulting mRNA accumulates RNA-binding proteins involved in splicing
Phu Le Thanh et al.
Neuromuscular disorders : NMD, 27(4), 338-351 (2017-02-19)
Reports of aberrant distribution for some nuclear envelope proteins in cells expressing a few Emery-Dreifuss muscular dystrophy mutations raised the possibility that such protein redistribution could underlie pathology and/or be diagnostic. However, this disorder is linked to 8 different genes
Gavin S Wilkie et al.
Molecular & cellular proteomics : MCP, 10(1), M110-M110 (2010-09-30)
Nuclear envelopes from liver and a neuroblastoma cell line have previously been analyzed by proteomics; however, most diseases associated with the nuclear envelope affect muscle. To determine whether muscle has unique nuclear envelope proteins, rat skeletal muscle nuclear envelopes were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service