Skip to Content
Merck
All Photos(1)

Key Documents

N-037

Supelco

(±)-Norketamine-D4 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Deuterated Norketamine Hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D4H11Cl2NO
Molecular Weight:
264.18
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.[2H]c1c([2H])c([2H])c(c(Cl)c1[2H])C2(N)CCCCC2=O

InChI

1S/C12H14ClNO.ClH/c13-10-6-2-1-5-9(10)12(14)8-4-3-7-11(12)15;/h1-2,5-6H,3-4,7-8,14H2;1H/i1D,2D,5D,6D;

InChI key

CLPOJGPBUGCUKT-LZLIYGNZSA-N

General description

Norketamine is the most prevalent urinary metabolite of ketamine, a club drug and widely used anesthetic sold under trade names including Ketanest®, Ketaset® and Ketalar®. This stable-labeled internal standard is suitable for quantitation of norketamine levels in GC/MS or LC/MS applications in clinical toxicology, urine drug testing, forensic analysis, clinical or diagnostic testing and isotope dilution methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Ketalar is a registered trademark of Par Sterile Products, LLC
Ketanest is a registered trademark of Pfizer, Inc.
Ketaset is a registered trademark of Zoetis W LLC
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service