Skip to Content
Merck
All Photos(2)

Key Documents

855780C

Avanti

2S-18:1 Lyso PC

Avanti Research - A Croda Brand

Synonym(s):

3-hydroxy-2-oleoyl-sn-glycero-1-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H52NO7P
Molecular Weight:
521.67
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

chloroform solution

packaging

package of 1 × 1 mL

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

5 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)CO)=O

Packaging

5 mL Clear Glass Sealed Ampule (855780-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service