Skip to Content
Merck
All Photos(1)

Key Documents

790145P

Avanti

23:2 Diyne PE [DC(8,9)PE]

1,2-bis(10,12-tricosadiynoyl)-sn-glycero-3-phosphoethanolamine, powder

Synonym(s):

DC(8,9)PE; DC8,9PE; DTPE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H86NO8P
CAS Number:
Molecular Weight:
872.20
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 200 mg (790145P-200mg)
pkg of 1 × 25 mg (790145P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 790145P

shipped in

dry ice

storage temp.

−20°C

Application

23:2 Diyne PE [DC(8,9)PE] or 1,2-bis(10,12-tricosadiynoyl)-sn-glycero-3-phosphoethanolamine has been used to form functional hybrid-liposomes.[1] It has also been used to modify single-walled carbon nanotubes (SWNTs).[2]

Biochem/physiol Actions

23:2 Diyne PE [DC(8,9)PE] or 1,2-bis(10,12-tricosadiynoyl)-sn-glycero-3-phosphoethanolamine is a photo-polymerizable lipid.[1] It forms intermolecular cross-linking through the diacetylenic groups. Thus, preferred for the formation of hybrid-liposomes.[1]

Packaging

20 mL Amber Glass Screw Cap Vial (790145P-200mg)
5 mL Amber Glass Screw Cap Vial (790145P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susan M Daly et al.
Langmuir : the ACS journal of surfaces and colloids, 22(3), 1215-1222 (2006-01-25)
We formed monolayers and black lipid membranes (BLMs) of photopolymerizable lipids mixed with the channel-forming protein gramicidin A to evaluate their miscibility and the potential for improved stability of the BLM scaffold through polymerization. Analyses of surface pressure vs area
Grzegorz Godlewski et al.
The Journal of pharmacology and experimental therapeutics, 328(1), 351-361 (2008-10-17)
The novel endocannabinoid-like lipid N-arachidonoyl L-serine (ARA-S) causes vasodilation through both endothelium-dependent and -independent mechanisms. We have analyzed the vasorelaxant effect of ARA-S in isolated vascular preparations and its effects on Ca(2+)-activated K(+) currents in human embryonic kidney cells stably
S Punnamaraju et al.
Langmuir : the ACS journal of surfaces and colloids, 28(20), 7657-7664 (2012-05-03)
A combination of nonpolymerizable phospholipids (DPPC or DPhPC) and a smaller amount of cross-linking photopolymerizable phospholipids (23:2 DiynePC) is incorporated in an unsupported artificial lipid bilayer formed using the droplet interface bilayer (DIB) approach. The DIB is formed by contacting
Peng He et al.
Biomacromolecules, 6(5), 2455-2457 (2005-09-13)
These studies show that single-walled carbon nanotubes (SWNTs) can be effectively modified using phospholipids. Using a simple surface modification of SWNTs, followed by deposition of 1,2-bis(10,12-tricosadiynoyl)-sn-glycero-3-phosphoethanolamine (DCPE) phosphipid, results in stable water-dispersible SWNTs with highly uniform thickness.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service