Skip to Content
Merck
All Photos(1)

Key Documents

750332P

Avanti

14:0 Cardiolipin (sodium salt)

1′,3′-bis[1,2-dimyristoyl-sn-glycero-3-phospho]-glycerol (sodium salt), powder

Synonym(s):

1,1′,2,2′-tetratetradecanoyl cardiolipin (sodium salt); CL(1′-[14:0/14:0],3′-[14:0/14:0])

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C65H124Na2O17P2
CAS Number:
Molecular Weight:
1285.60
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 g (750332P-1g)
pkg of 1 × 200 mg (750332P-200mg)
pkg of 1 × 25 mg (750332P-25mg)
pkg of 1 × 500 mg (750332P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 750332P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

Application

14:0 Cardiolipin (sodium salt) has been used:
  • as an internal standard for the quantitative analysis of cardiolipin (18:2)4 or CL (18:2)4 in human leukocytes and mouse mitochondria
  • for chemiluminescent assay
  • for the preparation of liposomes

Packaging

20 mL Clear Glass Screw Cap Vial (750332P-1g)
20 mL Clear Glass Screw Cap Vial (750332P-500mg)
5 mL Amber Glass Screw Cap Vial (750332P-200mg)
5 mL Amber Glass Screw Cap Vial (750332P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A rapid and sensitive LC-MS/MS method for quantitative analysis of cardiolipin (18: 2) 4 in human leukocytes and mouse skeletal muscles
Xu G, et al.
Journal of pharmaceutics, 386-394 (2018)
Specific interaction of the human mitochondrial uncoupling protein 1 with free long-chain fatty acid
Zhao L, et al.
Structure (2017)
Evaluation of cytochrome c affinity to anionic phospholipids by means of surface plasmon resonance
Stepanov G, et al.
The FEBS journal (2009)
Yuchen Sun et al.
Metabolites, 9(11) (2019-11-07)
Extracellular vesicles (EVs) consist of lipid bilayers, occur in various biofluids, and are invaluable in biomarker screening. Liquid chromatography coupled with high-resolution mass spectrometry (LC-MS) was recently used to study comprehensive EV lipid profiles in vitro. The aim of this
Julia Muenzner et al.
The journal of physical chemistry. B, 117(42), 12878-12886 (2013-05-30)
Interactions of cytochrome c (cyt c) with a unique mitochondrial glycerophospholipid cardiolipin (CL) are relevant for the protein's function in oxidative phosphorylation and apoptosis. Binding to CL-containing membranes promotes cyt c unfolding and dramatically enhances the protein's peroxidase activity, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service