Key Documents
Z742593
Accuris SmartDoc
2.0 Imaging System with blue light illumination base, AC/DC input 230 V AC, UK plug
Synonym(s):
gel documentation equipment, transilluminator system
About This Item
UNSPSC Code:
41106000
NACRES:
NB.24
feature
12V DC power supply included
High Intensity Blue LED; Orange PMMA photo filter and view cover
includes gel prep/cutting platform, blue light illumination base, orange filter cover, imaging enclosure and orange photo filter
packaging
pkg of 1
manufacturer/tradename
Benchmark Scientific E5001-SDB-E -UK       
W × D × H
23 cm × 19 cm × 22 cm, 2.6lbs (1.2kg) weight
compatibility
smartphones and tablets with camera
General description
Introducing the SmartDoc 2.0. This complete system has been developed for safely viewing nucleic acids in agarose gels and also for capturing quality digital images with a smart phone camera. Blue light is used instead of UV, which is safer for lab personnel and will not damage DNA samples. The illumination base emits an intense blue light with a peak output of 460nm for optimum excitation of nucleic acids stained with safe green dyes (SYBR Green, SmartGlow, GelStar, GelGreen, etc.) The orange cover fits on top of the illumination base and acts as a filter to absorb blue light, allowing visualization of the fluorescing samples. The cover can also be set at an angle for gel access and cutting out DNA bands for further study. The SmartDoc imaging enclosure includes an orange photo filter for photography of the gels with a smartphone. An extending adapter can be inserted for full dimension imaging of gels up to 15X15 cm or removed for close up pictures. The enclosure can also be used directly on any other existing UV or blue light trans illuminator. Additional filter are available for imaging gels on a UV trans illuminator.
Features:
  • Ideal for common safe stains (SmartGlow, SYBR Green, GelGreen, etc)
  • Visible blue light will not damage DNA
  • Quality gel imaging with any smart phone
  • Gel access for DNA band cutting
  • Imaging enclosure can also be used on existing trans illuminators
  • Maximum gel size: 15X15 cm
Legal Information
Accuris is a trademark of Benchmark Scientific, Inc.
SmartDoc is a trademark of Benchmark Scientific, Inc.