Key Documents
Z255858
Sigma-Aldrich
Latex rubber laboratory tubing
I.D. × O.D. 1/4 in. × 5/8 in.
About This Item
UNSPSC Code:
41105108
NACRES:
NB.51
packaging
pkg of 50 feet
I.D. × O.D.
1/4 in. × 5/8 in.
wall size
3/16 in.
General description
High-purity, seamless, amber tubing for general-purpose use with liquids and gases. Durometer: 35 ±5. Talc-free.