Key Documents
HAM75807
Hamilton® GC septa
PTFE/silicone (high temp), pack of 12 ea
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
Pricing and availability is not currently available.
material
PTFE/silicone (high temp)
yellow septa
packaging
pack of 12 ea
parameter
212 °C max. temp.
diam. × thickness
11.0 mm × 3.18 mm
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)