Skip to Content
Merck
All Photos(2)

Key Documents

926841

Sigma-Aldrich

1,4-Bis[4-(3-acryloyloxypropyloxy) benzoyloxy]-2-methylbenzene

greener alternative

≥97%

Synonym(s):

2-Methyl-1,4-phenylene bis(4-(3-(acryloyloxy)propoxy)benzoate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H32O10
CAS Number:
Molecular Weight:
588.60
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21

description

mesophase behaviour: Cr 64 N126I

Quality Level

Assay

≥97%

form

solid

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

white to off-white

greener alternative category

storage temp.

2-8°C

SMILES string

O(CCCOC(=O)C=C)c1ccc(cc1)C(=O)Oc2c(cc(cc2)OC(=O)c3ccc(cc3)OCCCOC(=O)C=C)C

InChI

1S/C33H32O10/c1-4-30(34)40-20-6-18-38-26-12-8-24(9-13-26)32(36)42-28-16-17-29(23(3)22-28)43-33(37)25-10-14-27(15-11-25)39-19-7-21-41-31(35)5-2/h4-5,8-17,22H,1-2,6-7,18-21H2,3H3

InChI key

ISSYGWIDLYOJEN-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to enabling category of greener alternatives and has been enhanced for energy efficiency. 4D printing basically produces zero waste. It holds the promise in the fight against climate change and manufacturing waste. Click here  for more information.

Application

1,4-Bis[4-(3-acryloyloxypropyloxy) benzoyloxy]-2-methylbenzene is a liquid-crystalline elastomer (LCE) monomer that has been explored as photonically , chemically or thermally fueled LCE actuator (LCEA) precursor for use in:
  • soft robotics, such as microscopic walkers , and self-healable actuators ;
  • 4d printing ;
  • amplifier reflectors for organic-inorganic perovskite solar-cells or for secure authentication .
  • optical sensors for medical applications .
This product can be used in Material Jetting or Vat polymerization 3D printing techniques.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuyun Liu et al.
Advanced materials (Deerfield Beach, Fla.), 29(9) (2016-12-23)
Azobenzene-containing cross-linked liquid crystal polymer films without hydrophilic groups exhibit dual-responsivity to humidity and UV light. The films realize not only a series of large and sophisticated contactless motions by utilizing moisture, including an inchworm walk, and tumbling locomotion, but
Enhanced Amplified Spontaneous Emission in Perovskites Using a Flexible Cholesteric Liquid Crystal Reflector.
Stranks S, et al.
Nano Letters, 15(8), 4935-4941 (2015)
Molecularly-Engineered, 4D-Printed Liquid Crystal Elastomer Actuators.
Saed M O, et al.
Advances in Functional Materials, 29, 1806412-1806412 (2019)
Yong Geng et al.
Scientific reports, 6, 26840-26840 (2016-05-28)
Monodisperse cholesteric liquid crystal microspheres exhibit spherically symmetric Bragg reflection, generating, via photonic cross communication, dynamically tuneable multi-coloured patterns. These patterns, uniquely defined by the particular sphere arrangement, could render cholesteric microspheres very useful in countless security applications, as tags
Samuel D Stranks et al.
Nano letters, 15(8), 4935-4941 (2015-05-20)
Organic-inorganic perovskites are highly promising solar cell materials with laboratory-based power conversion efficiencies already matching those of established thin film technologies. Their exceptional photovoltaic performance is in part attributed to the presence of efficient radiative recombination pathways, thereby opening up

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service