Skip to Content
Merck
All Photos(1)

Key Documents

612626

Supelco

Nicotine solution

NMR reference standard, 20 mg/mL in acetone-d6 (99.9 atom % D), TMS 0.05 %, NMR tube size 5 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2
Molecular Weight:
162.23
UNSPSC Code:
12142201
NACRES:
NA.24

grade

NMR reference standard
analytical standard

composition

TMS, 0.05%

concentration

20 mg/mL in acetone-d6 (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

5 mm × 8 in.

format

single component solution

Looking for similar products? Visit Product Comparison Guide

General description

NMR reference standard (20 mg/mL in acetone-d6 (99.9 atom % D), TMS 0.05%) eliminates the sample preparation uncertainty when evaluating instrument performance and assures proper instrument performance in quantitative applications. These NMR reference standards allow the users to monitor key operational parameters such as pw90, sensitivity, resolution, and line shape.

Quantity

5 mm O.D. tube contains 0.700 mL.
Each pack contains 10 tubes.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-2.2 °F

Flash Point(C)

-19 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service