Skip to Content
Merck
All Photos(4)

Key Documents

606022

Sigma-Aldrich

Sodium L-lactate-1-13C solution

greener alternative

45-55 % (w/w) in H2O, ≥99 atom % 13C, ≥98% (CP)

Synonym(s):

L-Lactic acid-1-13C, sodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)13CO2Na
CAS Number:
Molecular Weight:
113.05
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

description

Packaged as gram of solution, 45-55%(w/w)

isotopic purity

≥99 atom % 13C

Assay

≥98% (CP)

optical activity

[α]25/D −12°, c = 1 in H2O

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

concentration

45-55 % (w/w) in H2O

technique(s)

mass spectrometry (MS): suitable

mp

163-165 °C (lit.)

greener alternative category

mass shift

M+1

SMILES string

[Na+].C[C@H](O)[13C]([O-])=O

InChI

1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m0./s1/i3+1;

InChI key

NGSFWBMYFKHRBD-FWGDTYFGSA-M

Related Categories

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Safer Solvents and Auxiliaries” and “Design for Energy Efficiency”. Click here to view its DOZN scorecard.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service