Skip to Content
Merck
All Photos(1)

Key Documents

381829

Sigma-Aldrich

Diphosphoryl chloride

97%

Synonym(s):

Diphosphoric tetrachloride, Pyrophosphoric tetrachloride, Pyrophosphoryl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2P(O)OP(O)Cl2
CAS Number:
Molecular Weight:
251.76
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.475 (lit.)

bp

90 °C/12 mmHg (lit.)

density

1.82 g/mL at 25 °C (lit.)

SMILES string

ClP(Cl)(=O)OP(Cl)(Cl)=O

InChI

1S/Cl4O3P2/c1-8(2,5)7-9(3,4)6

InChI key

CNTIXUGILVWVHR-UHFFFAOYSA-N

Related Categories

Application

Useful reagent for the Vilsmeier formylation and glyoxylation reactions of nucleophilic aromatic compounds.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 49, 4015-4015 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service