Skip to Content
Merck
All Photos(1)

Key Documents

255343

Sigma-Aldrich

Isopropyl bromoacetate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CO2CH(CH3)2
CAS Number:
Molecular Weight:
181.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.444 (lit.)

bp

59-61 °C/10 mmHg (lit.)

density

1.399 g/mL at 25 °C (lit.)

SMILES string

CC(C)OC(=O)CBr

InChI

1S/C5H9BrO2/c1-4(2)8-5(7)3-6/h4H,3H2,1-2H3

InChI key

JCWLEWKPXYZHGQ-UHFFFAOYSA-N

Related Categories

Application

Isopropyl bromoacetate has been used in the synthesis of biaryl sulphonamide derivatives.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jon J Hangeland et al.
Bioorganic & medicinal chemistry letters, 18(2), 474-478 (2007-12-28)
T-type calcium channel antagonists were designed using a protocol involving the program SPROUT and constrained by a ComFA-based pharmacophore model. Scaffolds generated by SPROUT were evaluated based on their ability to be translated into structures that were synthetically tractable. From

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service