Skip to Content
Merck
All Photos(1)

Documents

238902

Sigma-Aldrich

2-(Trimethylsilyl)ethoxymethyl chloride

technical grade

Synonym(s):

(2-Chloromethoxyethyl)trimethylsilane, Chloromethyl 2-trimethylsilylethyl ether, SEM-Cl, SEM-chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCH2CH2OCH2Cl
CAS Number:
Molecular Weight:
166.72
Beilstein:
3587289
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

form

liquid

refractive index

n20/D 1.435 (lit.)

bp

170-172 °C (lit.)
57-59 °C/8 mmHg (lit.)

density

0.942 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

C[Si](C)(C)CCOCCl

InChI

1S/C6H15ClOSi/c1-9(2,3)5-4-8-6-7/h4-6H2,1-3H3

InChI key

BPXKZEMBEZGUAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Learn More at the Professor and Product Portal of Professor Bruce Lipshutz.

Application

Phenol protecting group in the synthesis of laterifluorones.
Used to prepare a SEM-ether which was, in turn, removed with BBr3 and cyclized to a benzo-oxapane.

Features and Benefits

Convenient hydroxyl-protecting reagent. SEM-ethers are stable over a wide pH range, and can be selectively cleaved with fluoride ion under mild aprotic conditions.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 36, 1683-1683 (1995)
K C Nicolaou et al.
Journal of the American Chemical Society, 126(17), 5493-5501 (2004-04-29)
The total synthesis of 1-O-methyllateriflorone (2) is described. The construction of the cage-like domain of the molecule involved a biomimetic Claisen/Diels-Alder cascade, whereas the novel spiroxalactone framework was generated by an intramolecular Michael reaction within precursor 16a involving the carboxylate
The Journal of Organic Chemistry, 55, 5180-5180 (1990)
Tetrahedron Letters, 47, 5909-5909 (2006)

Related Content

Prof. Bruce Lipshutz and co-workers have developed designer surfactants to allow several classes of transformations (e.g. Suzuki-Miyaura, Olefin Metathesis, 1,4-Addition to Enones, etc.) to be performed in water.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service