Skip to Content
Merck
All Photos(1)

Documents

238333

Sigma-Aldrich

1-Bromopentadecane

97%

Synonym(s):

Pentadecyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14Br
CAS Number:
Molecular Weight:
291.31
Beilstein:
1744553
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

10.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.461 (lit.)

bp

159-160 °C/5 mmHg (lit.)

mp

17-19 °C (lit.)

density

1.005 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCBr

InChI

1S/C15H31Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16/h2-15H2,1H3

InChI key

JKOTZBXSNOGCIF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Incorporation of 1-bromopentadecane into mixed monolayers of deuterated palmitic acid (dPA) and deuterated stearic acid (dSA) at the air-water interface has been investigated.

Application

1-Bromopentadecane has been used as regent in the synthesis of (S)-eicos-(4E)-en-1-yn-3-ol, a bioactive component of the marine sponge Cribrochalina vasculum.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntheses of two enantiomers of eicos-(4E)-en-1-yn-3-ol, a bioactive component of the marine sponge Cribrochalina vasculum.
Lu W, et al.
Tetrahedron, 55(15), 4649-4654 (1999)
M Roxana Sierra-Hernández et al.
Langmuir : the ACS journal of surfaces and colloids, 26(24), 18806-18816 (2010-12-02)
Mixed monolayers of deuterated palmitic acid C(15)D(31)COOH (dPA) and deuterated stearic acid C(17)D(35)COOH (dSA) with 1-bromoalkanes of different alkyl chain length (C(4) to C(16)) at the air-water interface were investigated. Alkanes and 1-chlorohexadecane ClC(16)H(33) (ClHex) were also studied to compare
Fabien Lacaille et al.
PloS one, 2(7), e661-e661 (2007-08-22)
Sexual behavior requires animals to distinguish between the sexes and to respond appropriately to each of them. In Drosophila melanogaster, as in many insects, cuticular hydrocarbons are thought to be involved in sex recognition and in mating behavior, but there

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service