Skip to Content
Merck
All Photos(1)

Key Documents

138673

Sigma-Aldrich

4-Bromophenylacetic acid

4-Bromophenylacetic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH2CO2H
CAS Number:
Molecular Weight:
215.04
Beilstein:
972146
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

powder

mp

114-117 °C (lit.)

solubility

ethanol: soluble 5%, clear, colorless to light yellow

functional group

bromo
carboxylic acid

SMILES string

OC(=O)Cc1ccc(Br)cc1

InChI

1S/C8H7BrO2/c9-7-3-1-6(2-4-7)5-8(10)11/h1-4H,5H2,(H,10,11)

InChI key

QOWSWEBLNVACCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Bromophenylacetic acid was used as standard in the extraction of nonylphenol polyethoxy carboxylates with 1-4 ethoxy groups from sludge samples[1].

Biochem/physiol Actions

4-Bromophenylacetic acid is a growth inhibitory substance[2]. It causes the depolarization effect on the transmembrane potential difference of tobacco protoplasts isolated from two genotypes differing in their sensitivity to auxins[2].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Ephritikhine et al.
Plant physiology, 83(4), 801-804 (1987-04-01)
The effects of 1-naphthaleneacetic acid (NAA) and other auxin analogs on the transmembrane potential difference (Em) were compared on tobacco protoplasts isolated from two genotypes differing in their sensitivity to auxins. For both types, NAA modifies Em by inducing at
Subcritical (hot) water/ethanol extraction of nonylphenol polyethoxy carboxylates from industrial and municipal sludges.
Field JA and Reed RL.
Environmental Science & Technology, 33(16), 2782-2787 (1999)

Questions

Reviews

Active Filters

  1. INDIA
    • Review 1
    • Votes 0
    3 out of 5 stars.

    4-bromophenylacetic acid

    Please correct structure of 4-bromophenylacetic acid, it is wrong

    Helpful?

    1. Response from MilliporeSigma:

      Thank you for your review. We are sorry to hear that your experience did not match your product expectations. We encourage customers who experience problems with our products to contact us for additional technical support. Please visit https://www.sigmaaldrich.com/US/en/support/customer-support to submit a Product Technical Inquiry.

  2. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service