Merck
HPLC Analysis of PAHs in Water on SUPELCOSIL LC-PAH after SPME using 100 μm PDMS Fiber
HPLC Analysis of PAHs in Water on SUPELCOSIL™ LC-PAH after SPME using 100 μm PDMS Fiber application for SPME, application for HPLC
Materials
analytical column
Product No.
Description
Pricing
SUPELCOSIL LC-PAH HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
SPME fiber
Product No.
Description
Pricing
SPME Fiber Assembly
100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
CONDITIONS
sample/matrix
5 μL PAH mix (Cat. No. 4-8743) in 5 mL water
SPME fiber
polydimethylsiloxane, 100 μm (57301)
extraction
immersion, 30 min (rapid stirring)
desorption process
static, 200 μL acetonitrile:water, 40:60, 2 min
column
SUPELCOSIL LC-PAH, 15 cm × 4.6 mm I.D., 5 μm particles (58318)
gradient
0 min - 50% A; 5 min - 50% A; 30 min - 100% A; 45 min - 100% A
flow rate
0-2 min: 0.2 mL/min 2-45 min: 1.0 mL/min
mobile phase
[A] acetonitrile:[B] water
detector
UV, 254 nm
Description
Legal Information
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC