Merck
  • Disruption of Rab8a and Rab11a causes formation of basolateral microvilli in neonatal enteropathy.
Disruption of Rab8a and Rab11a causes formation of basolateral microvilli in neonatal enteropathy.
Journal of cell science (2017-06-10)
Qiang Feng, Edward M Bonder, Amy C Engevik, Lanjing Zhang, Matthew J Tyska, James R Goldenring, Nan Gao
ABSTRACT
Misplaced formation of microvilli to basolateral domains and intracellular inclusions in enterocytes are pathognomonic features in congenital enteropathy associated with mutation of the apical plasma membrane receptor syntaxin 3 (STX3). Although the demonstrated binding of Myo5b to the Rab8a and Rab11a small GTPases in vitro implicates cytoskeleton-dependent membrane sorting, the mechanisms underlying the microvillar location defect remain unclear. By selective or combinatory disruption of Rab8a and Rab11a membrane traffic in vivo, we demonstrate that transport of distinct cargo to the apical brush border rely on either individual or both Rab regulators, whereas certain basolateral cargos are redundantly transported by both factors. Enterocyte-specific Rab8a and Rab11a double-knockout mouse neonates showed immediate postnatal lethality and more severe enteropathy than single knockouts, with extensive formation of microvilli along basolateral surfaces. Notably, following an inducible Rab11a deletion from neonatal enterocytes, basolateral microvilli were induced within 3ā€…days. These data identify a potentially important and distinct mechanism for a characteristic microvillus defect exhibited by enterocytes of patients with neonatal enteropathy.
Sigma-Aldrich
BIO, ≥98% (HPLC)
Sigma-Aldrich
Anti-PACSIN2 antibody produced in rabbit, Prestige AntibodiesĀ® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution