Merck
  • Design, Synthesis, Spectral Analysis, In Vitro Anticancer Evaluation and Molecular Docking Studies of Some Fluorescent 4-Amino-2, 3-Dimethyl-1-Phenyl-3-Pyrazolin-5-One, Ampyrone Derivatives.
Design, Synthesis, Spectral Analysis, In Vitro Anticancer Evaluation and Molecular Docking Studies of Some Fluorescent 4-Amino-2, 3-Dimethyl-1-Phenyl-3-Pyrazolin-5-One, Ampyrone Derivatives.
Interdisciplinary sciences, computational life sciences (2016-01-05)
D Premnath, Israel V M V Enoch, P Mosae Selvakumar, M Indiraleka, J Jannet Vennila
ABSTRACT
The commenced work deals with the synthesis, characterization and evaluation of biological activities of 4-amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one. The synthesis was done by the condensation of aromatic acid chlorides with 4-aminoantipyrine. The structures of synthesized derivatives were elucidated using IR, Mass,
Sigma-Aldrich
3,5-Bis(trifluoromethyl)benzoyl chloride, 97%
Sigma-Aldrich
1,3-Propylene sulfite, 99%