Merck
  • Cleavage of neuronal synaptosomal-associated protein of 25 kDa by exogenous matrix metalloproteinase-7.
Cleavage of neuronal synaptosomal-associated protein of 25 kDa by exogenous matrix metalloproteinase-7.
Journal of neurochemistry (2007-05-03)
Arek Szklarczyk, George Oyler, Ron McKay, Charles Gerfen, Katherine Conant
ABSTRACT
Matrix metalloproteinases (MMPs) belong to a family of zinc dependent enzymes best studied for their role in cancer and inflammation. Though MMPs typically target extracellular proteins, here we show that MMP-7, an MMP family member which lacks a C-terminal hemopexin-like domain, can cleave an intraneuronal protein that is critical to vesicular fusion and neurotransmitter release, synaptosomal-associated protein of 25 kDa (SNAP-25). Western blot analysis using an N-terminal specific antibody on extracts from cultured neurons suggests that cleavage occurs towards the C-terminal portion of SNAP 25. Additional studies with recombinant SNAP-25 demonstrate that cleavage occurs at amino acid 129. The ability of MMP-7 to cleave SNAP-25 is diminished by chlorpromazine and phenylarsine oxide, inhibitors of clathrin dependent endocytosis. Together, these results imply that exogenous MMP-7 can access an intraneuronal substrate and suggest that additional studies may be warranted to determine if SNAP function is impaired with brain inflammation.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Anti-SNAP-25 Antibody, Chemicon®, from goat
Sigma-Aldrich
MMP-9, Active, Human, Recombinant
Sigma-Aldrich
GM 6001, GM 6001, CAS 142880-36-2, is a potent, cell-permeable, broad-spectrum inhibitor of matrix metalloproteinases.