Merck
  • Measuring cell surface area and deformability of individual human red blood cells over blood storage using quantitative phase imaging.
Measuring cell surface area and deformability of individual human red blood cells over blood storage using quantitative phase imaging.
Scientific reports (2016-10-05)
HyunJoo Park, SangYun Lee, Misuk Ji, Kyoohyun Kim, YongHak Son, Seongsoo Jang, YongKeun Park
ABSTRACT
The functionality and viability of stored human red blood cells (RBCs) is an important clinical issue in transfusions. To systematically investigate changes in stored whole blood, the hematological properties of individual RBCs were quantified in blood samples stored for various periods with and without a preservation solution called citrate phosphate dextrose adenine-1 (CPDA-1). With 3-D quantitative phase imaging techniques, the optical measurements for 3-D refractive index (RI) distributions and membrane fluctuations were done at the individual cell level. From the optical measurements, the morphological (volume, surface area and sphericity), biochemical (hemoglobin content and concentration), and mechanical parameters (dynamic membrane fluctuation) were simultaneously quantified to investigate the functionalities and progressive alterations of stored RBCs. Our results show that stored RBCs without CPDA-1 had a dramatic morphological transformation from discocytes to spherocytes within two weeks which was accompanied by significant decreases in cell deformability and cell surface area, and increases in sphericity. However, the stored RBCs with CPDA-1 maintained their morphology and deformability for up to 6 weeks.
Sigma-Aldrich
Citrate-phosphate-dextrose solution with adenine
Pricing and availability is not currently available.