Merck
  • Progesterone regulates HLA-G gene expression through a novel progesterone response element.
Progesterone regulates HLA-G gene expression through a novel progesterone response element.
Human reproduction (Oxford, England) (2006-05-11)
Shang-mian Yie, Rong Xiao, Clifford L Librach
ABSTRACT
We have previously demonstrated that progesterone has a stimulatory effect on HLA-G gene expression. Because this effect was abolished by the anti-progestin, RU486, we hypothesize that this effect is through receptor-mediated up-regulation of the HLA-G gene. The objective of this study was to explore the molecular mechanisms of this effect. The transient transfection of a chloramphenicol acetyltransferase (CAT) construct containing a fragment of the HLA-G gene promoter into the JEG-3 choriocarcinoma cell line was performed. An electrophoretic mobility shift assay (EMSA) and a DNA fragment-binding enzyme-linked immunosorbent assay (ELISA) were carried out to locate a specific progesterone response element (PRE) in the HLA-G gene promoter region. Progesterone treatment of JEG-3 cells transfected with the HLA-G gene promoter-CAT construct resulted in an increase of CAT synthesis, whereas RU486 blocked this transcriptional activation. A novel PRE-binding site sequence, with 60% homology to that of wild-type mouse mammary tumour virus (MMTV) PRE, was discovered in this region. The effect of progesterone on HLA-G gene expression is through progesterone receptor (PR) activation, followed by binding to a novel PRE in the HLA-G promoter region. Therefore, one of the mechanisms of immunomodulation by progesterone during pregnancy may be through the regulation of HLA-G gene expression via this novel PRE.
Product Description
Roche
Fluorescein-12-dUTP, ≥85% (HPLC), suitable for ELISA, suitable for hybridization, solution
Pricing and availability is not currently available.