Merck
  • Single-tube linear DNA amplification for genome-wide studies using a few thousand cells.
Single-tube linear DNA amplification for genome-wide studies using a few thousand cells.
Nature protocols (2012-01-28)
Pattabhiraman Shankaranarayanan, Marco-Antonio Mendoza-Parra, Wouter van Gool, Luisa M Trindade, Hinrich Gronemeyer
ABSTRACT
Linear amplification of DNA (LinDA) by T7 polymerase is a versatile and robust method for generating sufficient amounts of DNA for genome-wide studies with minute amounts of cells. LinDA can be coupled to a great number of global profiling technologies. Indeed, chromatin immunoprecipitation coupled to massive parallel sequencing (ChIP-seq) has been achieved for transcription factors and epigenetic modification of chromatin histones with 1,000 to 5,000 cells. LinDA largely simplifies reChIP-seq experiments to monitor co-binding at chromatin target sites. The single-tube design of LinDA is ideal for handling ultrasmall amounts of DNA (<30 pg) and is compatible with automation. The actual hands-on working time is less than 6 h with one overnight reaction. The present protocol describes all materials and critical steps, and provides examples and controls for LinDA. Applications of LinDA for genome-wide analyses of biobank samples and for the study of chromatin conformation and nuclear architecture are in progress.
Product Description
Roche
Taq DNA Polymerase, 5 U/μl, optimum pH ~9.0 (20 °C), optimum reaction temp. 72 °C
Pricing and availability is not currently available.
Sigma-Aldrich
GenElute Mammalian Total RNA Miniprep Kit, sufficient for 70 purifications
Pricing and availability is not currently available.