Merck
  • Action of Lovastatin (Mevinolin) on an in vitro model of angiogenesis and its co-culture with malignant melanoma cell lines.
Action of Lovastatin (Mevinolin) on an in vitro model of angiogenesis and its co-culture with malignant melanoma cell lines.
Cancer cell international (2006-04-01)
Ivan Depasquale, Denys N Wheatley
ABSTRACT
Lovastatin and other statins may reduce the development of melanomas. The effects on melanoma cells and their ability to enhance angiogenesis in a co-culture system presented an opportunity to assess whether Lovastatin act on melanoma cells, HUVEC or both types of cells. Direct effects of co-culturing two different malignant melanoma cells (A375 and G361) on the process of angiogenesis in vitro was studied with our angiogenesis model, based on human dermal fibroblasts and human umbilical vein endothelial cells (HUVEC). Co-cultures were set up using "sland" and "dispersed seeding" techniques. A statistically significant increase in tubule formation in both cases was observed compared to controls. The effects of doses equivalent to therapeutic concentrations of Lovastatin were analysed. The drug inhibited the growth of all cell types, induced apoptosis, and markedly reduced the formation of tubules in the angiogenesis model at low concentrations. Its action was successfully reversed by the introduction of geranylgeranyl pyrophosphate. Lovastatin can reduce both tumour (melanoma) cell growth, and the angiogenic activity of these cells in co-cultures using an established 2-dimensional model angiogenesis system beyond that which would be seen by reduced proliferation alone.
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
Sigma-Aldrich
Anti-MCAM Antibody, clone P1H12, clone P1H12, Chemicon®, from mouse