Merck
  • Broadening of polymer chromatographic signals: Analysis, quantification and correction through effective diffusion coefficients.
Broadening of polymer chromatographic signals: Analysis, quantification and correction through effective diffusion coefficients.
Journal of chromatography. A (2015-07-15)
Inmaculada Suárez, Baudilio Coto
ABSTRACT
Average molecular weights and polydispersity indexes are some of the most important parameters considered in the polymer characterization. Usually, gel permeation chromatography (GPC) and multi angle light scattering (MALS) are used for this determination, but GPC values are overestimated due to the dispersion introduced by the column separation. Several procedures were proposed to correct such effect usually involving more complex calibration processes. In this work, a new method of calculation has been considered including diffusion effects. An equation for the concentration profile due to diffusion effects along the GPC column was considered to be a Fickian function and polystyrene narrow standards were used to determine effective diffusion coefficients. The molecular weight distribution function of mono and poly disperse polymers was interpreted as a sum of several Fickian functions representing a sample formed by only few kind of polymer chains with specific molecular weight and diffusion coefficient. Proposed model accurately fit the concentration profile along the whole elution time range as checked by the computed standard deviation. Molecular weights obtained by this new method are similar to those obtained by MALS or traditional GPC while polydispersity index values are intermediate between those obtained by the traditional GPC combined to Universal Calibration method and the MALS method. Values for Pearson and Lin coefficients shows improvement in the correlation of polydispersity index values determined by GPC and MALS methods when diffusion coefficients and new methods are used.
Sigma-Aldrich
Triisobutylaluminum solution, 1.0 M in hexanes
Sigma-Aldrich
Toluene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Toluene, JIS 300, for residue analysis, ≥99.8%
Sigma-Aldrich
Toluene, JIS 1000, for residue analysis, ≥99.8%
Sigma-Aldrich
Toluene, JIS special grade, ≥99.5%
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Toluene, HPLC grade, 99.8%