Merck
  • Determination of Toxic Metals in Little Cigar Tobacco with 'Triple Quad' ICP-MS.
Determination of Toxic Metals in Little Cigar Tobacco with 'Triple Quad' ICP-MS.
Journal of analytical toxicology (2015-03-01)
R Steven Pappas, Naudia Martone, Nathalie Gonzalez-Jimenez, Mark R Fresquez, Clifford H Watson
ABSTRACT
Smoking remains the leading cause of preventable death in the USA. Much of the focus on harmful and potentially harmful constituents (HPHCs) in tobacco products has been on cigarettes. Little cigars gained popularity over the last decade until tobacco taxes made cigarettes more expensive in the USA. Many little cigar brands are similar in size with cigarettes and may be smoked in a similar manner. Scant data are available on HPHC concentrations in little cigars, therefore we developed and applied a new analytical method to determine concentrations of 10 toxic metals in little cigar tobacco. The method utilizes 'triple quadrupole' ICP-MS. By optimizing octapole bias, energy discrimination and cell gas flow settings, we were able to accurately quantify a range of elements including those for which the cell gas reactions were endothermic. All standard modes (Single Quad No Gas, MS-MS NH3/He and MS-MS O2) were utilized for the quantitation of 10 toxic metals in little cigar tobacco, including uranium, which was added as an analyte in the new method. Because of the elimination of interfering ions at 'shifted analyte masses', detection limits were lower compared with a previous method. Tobacco selenium concentrations were below the limit of detection in the previous method, but the new technology made it possible to report all selenium concentrations.
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
Thulium Standard for ICP, TraceCERT®, 1000 mg/L Tm in nitric acid
Sigma-Aldrich
Nitric acid, 1 M
Sigma-Aldrich
Nitric acid, 0.1 M
Sigma-Aldrich
Nitric acid, SAJ first grade, 65.0-66.0%, density: 1.40
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, suitable for HPLC
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, JIS special grade, ≥99.5%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis