Merck
  • Increased Levels of Antinutritional and/or Defense Proteins Reduced the Protein Quality of a Disease-Resistant Soybean Cultivar.
Increased Levels of Antinutritional and/or Defense Proteins Reduced the Protein Quality of a Disease-Resistant Soybean Cultivar.
Nutrients (2015-07-25)
Daniele O B Sousa, Ana F U Carvalho, José Tadeu A Oliveira, Davi F Farias, Ivan Castelar, Henrique P Oliveira, Ilka M Vasconcelos
ABSTRACT
The biochemical and nutritional attributes of two soybean (Glycine max (L.) Merr.) cultivars, one susceptible (Seridó) and the other resistant (Seridó-RCH) to stem canker, were examined to assess whether the resistance to pathogens was related to levels of antinutritional and/or defense proteins in the plant and subsequently affected the nutritional quality. Lectin, urease, trypsin inhibitor, peroxidase and chitinase activities were higher in the resistant cultivar. Growing rats were fed with isocaloric and isoproteic diets prepared with defatted raw soybean meals. Those on the Seridó-RCH diet showed the worst performance in terms of protein quality indicators. Based on regression analysis, lectin, trypsin inhibitor, peroxidase and chitinase appear to be involved in the resistance trait but also in the poorer nutritional quality of Seridó-RCH. Thus, the development of cultivars for disease resistance may lead to higher concentrations of antinutritional compounds, affecting the quality of soybean seeds. Further research that includes the assessment of more cultivars/genotypes is needed.
Sigma-Aldrich
Guaiacol, SAJ first grade, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥95% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Nα-Benzoyl-L-arginine 4-nitroanilide hydrochloride, ≥99% (TLC), suitable for substrate for trypsin
Pricing and availability is not currently available.
Sigma-Aldrich
Trypsin from human pancreas, salt-free, lyophilized powder, vial of ≥1,000 BAEE units
Pricing and availability is not currently available.
Sigma-Aldrich
Nα-Benzoyl-L-arginine 4-nitroanilide hydrochloride, >98% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Guaiacol, oxidation indicator
Pricing and availability is not currently available.
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
N-Acetyl-D-glucosamine, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), lyophilized powder
Pricing and availability is not currently available.
Sigma-Aldrich
Laminarin from Laminaria digitata, polysaccharide substrate for laminarinase
Pricing and availability is not currently available.
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-1, ≥1,000,000 units/g solid
Pricing and availability is not currently available.
Sigma-Aldrich
Egg whites from chicken
Pricing and availability is not currently available.
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type C-3, powder, ≥600,000 units/g solid
Pricing and availability is not currently available.