Merck
  • Heterotrophic-autotrophic sequential system for reductive nitrate and perchlorate removal.
Heterotrophic-autotrophic sequential system for reductive nitrate and perchlorate removal.
Environmental technology (2015-06-24)
Deniz Ucar, Emine Ubay Cokgor, Erkan Sahinkaya
ABSTRACT
Nitrate and perchlorate were identified as significant water contaminants all over the world. This study aims at evaluating the performances of the heterotrophic-autotrophic sequential denitrification process for reductive nitrate and perchlorate removal from drinking water. The reduced nitrate concentration in the heterotrophic reactor increased with increasing methanol concentrations and the remaining nitrate/nitrite was further removed in the following autotrophic denitrifying process. The performances of the sequential process were studied under varying nitrate loads of [Formula: see text] at a fixed hydraulic retention time of 2 h. The C/N ratio in the heterotrophic reactor varied between 1.24 and 2.77 throughout the study. Nitrate and perchlorate reduced completely with maximum initial concentrations of [Formula: see text] and 1000 µg/L, respectively. The maximum denitrification rate for the heterotrophic reactor was [Formula: see text] when the bioreactor was fed with [Formula: see text] and 277 mg/L methanol. For the autotrophic reactor, the highest denitrification rate was [Formula: see text] in the first period when the heterotrophic reactor performance was low. Perchlorate reduction was initiated in the heterotrophic reactor, but completed in the following autotrophic process. Effluent sulphate concentration was below the drinking water standard level of 250 mg/L and pH was in the neutral level.
Sigma-Aldrich
Sodium perchlorate monohydrate, puriss. p.a., ACS reagent, ≥98.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium perchlorate monohydrate, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium nitrate, SAJ first grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium perchlorate hydrate, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium nitrate, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium nitrate, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium nitrate, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium nitrate, ReagentPlus®, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium nitrate, puriss., meets analytical specification of Ph Eur, BP, USP, FCC, E252, 99.0-100.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Pricing and availability is not currently available.