Merck
  • A method for quantification of volatile organic compounds in blood by SPME-GC-MS/MS with broader application: From non-occupational exposure population to exposure studies.
A method for quantification of volatile organic compounds in blood by SPME-GC-MS/MS with broader application: From non-occupational exposure population to exposure studies.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-05-13)
Rocio Aranda-Rodriguez, Ashley Cabecinha, Jeromy Harvie, Zhiyun Jin, Axelle Marchand, Robert Tardif, Andy Nong, Sami Haddad
ABSTRACT
Humans are continuously exposed to volatile organic compounds (VOCs) as these chemicals are ubiquitously present in most indoor and outdoor environments. In order to assess recent exposure to VOCs for population-based studies, VOCs are measured in the blood of participants. This work describes an improved method to detect 12 VOCs by head-space solid-phase microextraction gas chromatography coupled with isotope-dilution mass spectrometry in selected reaction monitoring mode (SPME-GC-MS/MS). This method was applied to the analysis of trihalomethanes, styrene, trichloroethylene, tetrachloroethylene and BTEX (benzene, toluene, ethylbenzene, m-xylene, p-xylene, o-xylene) in a population-based biomonitoring study (Canadian Health Measures Survey). The method showed good linearity (>0.990) in the range of 0.010-10μg/L and detection limits between 0.007 and 0.027μg/L, precision better than 25% and good accuracy (±25%) based on proficiency testing materials. Quality Control data among runs over a 7 month period showed %RSD between 14 and 25% at low levels (∼0.03μg/L) and between 9 and 23% at high levels (∼0.4μg/L). The method was modified to analyze samples from a pharmacokinetic study in which 5 healthy volunteers were exposed to single, binary and quaternary mixtures of CTEX (chloroform, ethylbenzene, toluene and m-xylene), thus the expected concentration in blood was 1 order of magnitude higher than those found in the general population. The method was modified by reducing the sample size (from 3g to 0.5g) and increasing the upper limit of the concentration range to 395μg/L. Good linearity was found in the range of 0.13-395μg/L for toluene and ethylbenzene and 0.20-609μg/L for m/p-xylene. Quality control data among runs over the period of the study (n=13) were found to vary between 7 and 25%.
Sigma-Aldrich
Trichloroethylene-13C2, 99 atom % 13C, 97% (CP)
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga, metal alloy fiber
Sigma-Aldrich
Benzene, JIS special grade, ≥99.5%
Sigma-Aldrich
Benzene, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Sodium fluoride, JIS special grade, ≥99.0%
Sigma-Aldrich
Benzene, SAJ first grade, ≥99.0%
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assortment kit 2, for use with autosampler, size 24 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, size 24 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 2, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 2, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, needle size 23 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 23 ga, StableFlex fiber, for use with autosampler
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
SPME StableFlex fiber assortment kit, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 23 ga
Supelco
SPME StableFlex fiber assortment kit, for use with manual holder
Supelco
SPME fiber assortment kit 4, for use with manual holder, size 24 ga
Supelco
SPME fiber assortment kit 1, for use with manual holder, needle size 24 ga
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Supelco
SPME fiber assortment kit 5, needle size 23 ga, for use with autosampler
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Styrene, SAJ first grade, ≥99.0%
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, metal alloy fiber, for use with manual holder