Merck
  • Nd(3+)-Sensitized Ho(3+) Single-Band Red Upconversion Luminescence in Core-Shell Nanoarchitecture.
Nd(3+)-Sensitized Ho(3+) Single-Band Red Upconversion Luminescence in Core-Shell Nanoarchitecture.
The journal of physical chemistry letters (2015-08-13)
Daqin Chen, Lu Liu, Ping Huang, Mingye Ding, Jiasong Zhong, Zhenguo Ji
ABSTRACT
A strategy to achieve 808 nm excited single-band red upconversion luminescence of Ho(3+) via the core-shell nanoarchitecture design was provided. Specifically, the synthesized Yb/Ho/Ce: NaGdF4@Yb/Nd: NaYF4 active-core@active-shell nanoparticles were evidenced to enable high-content doping of Nd(3+) (∼10 mol %) in the shell layer and, thus, markedly enhance red upconversion emission from Ho(3+) activators in the core with the assistance of spatially confined doping of Nd(3+) ions and efficient energy transfer of Nd(3+) → Yb(3+)(shell) → Yb(3+)(core) → Ho(3+). Importantly, introducing Ce(3+) into the core was beneficial to the competition of radiation transitions from the two intermediate excited states of Ho(3+): (5)S2,(5)F4 (green-emitting) and Ho(3+): (5)F5 (red-emitting), which induced great enhancement in the red to green intensity ratio and ultimately intense single-band red upconversion emission. We believe that this preliminary study will provide an important advance in developing luminescent markers suitable for biolabeling applications.
Sigma-Aldrich
Sodium hydroxide solution, 0.2 M
Sigma-Aldrich
Sodium hydroxide, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Sigma-Aldrich
Sodium hydroxide solution, 7 M
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Holmium(III) chloride, anhydrous, powder, 99.9% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Gadolinium(III) chloride solution, NMR reference standard, 0.1 mg/mL in D2O (99.9 atom % D), Methanol-13C 0.1 % (99 atom % 13C), water 1 %, NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Gadolinium(III) chloride solution, NMR reference standard, 0.1 mg/mL in D2O (99.9 atom % D), water 0.1 %, NMR tube size 6.5 mm × 8 in.
Sigma-Aldrich
1-Octadecene, ≥95.0% (GC)
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Neodymium(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Yttrium(III) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Cerium(III) chloride, AnhydroBeads, −10 mesh, 99.9%
Sigma-Aldrich
Gadolinium(III) chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
Yttrium(III) chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Cerium(III) chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis