Merck
  • Copper-catalyzed intermolecular carboetherification of unactivated alkenes by alkyl nitriles and alcohols.
Copper-catalyzed intermolecular carboetherification of unactivated alkenes by alkyl nitriles and alcohols.
Angewandte Chemie (International ed. in English) (2015-03-05)
Claire Chatalova-Sazepin, Qian Wang, Glenn M Sammis, Jieping Zhu
ABSTRACT
A three-component carboetherification of unactivated alkenes has been developed allowing the rapid building of complexity from simple starting materials. A wide range of α-substituted styrenes underwent smooth reactions with unactivated alkyl nitriles and alcohols to afford γ-alkoxy alkyl nitriles with concomitant generation of a quaternary carbon center. A radical clock experiment provided clear-cut evidence that the reaction proceeds through a tertiary alkyl radical intermediate.
Sigma-Aldrich
Benzophenone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Benzophenone, purified by sublimation, ≥99%
Sigma-Aldrich
Benzophenone, ReagentPlus®, 99%