Merck
  • Complex reference values for endocrine and special chemistry biomarkers across pediatric, adult, and geriatric ages: establishment of robust pediatric and adult reference intervals on the basis of the Canadian Health Measures Survey.
Complex reference values for endocrine and special chemistry biomarkers across pediatric, adult, and geriatric ages: establishment of robust pediatric and adult reference intervals on the basis of the Canadian Health Measures Survey.
Clinical chemistry (2015-06-06)
Khosrow Adeli, Victoria Higgins, Michelle Nieuwesteeg, Joshua E Raizman, Yunqi Chen, Suzy L Wong, David Blais
ABSTRACT
Defining laboratory biomarker reference values in a healthy population and understanding the fluctuations in biomarker concentrations throughout life and between sexes are critical to clinical interpretation of laboratory test results in different disease states. The Canadian Health Measures Survey (CHMS) has collected blood samples and health information from the Canadian household population. In collaboration with the Canadian Laboratory Initiative on Pediatric Reference Intervals (CALIPER), the data have been analyzed to determine reference value distributions and reference intervals for several endocrine and special chemistry biomarkers in pediatric, adult, and geriatric age groups. CHMS collected data and blood samples from thousands of community participants aged 3 to 79 years. We used serum samples to measure 13 immunoassay-based special chemistry and endocrine markers. We assessed reference value distributions and, after excluding outliers, calculated age- and sex-specific reference intervals, along with corresponding 90% CIs, according to CLSI C28-A3 guidelines. We observed fluctuations in biomarker reference values across the pediatric, adult, and geriatric age range, with stratification required on the basis of age for all analytes. Additional sex partitions were required for apolipoprotein AI, homocysteine, ferritin, and high sensitivity C-reactive protein. The unique collaboration between CALIPER and CHMS has enabled, for the first time, a detailed examination of the changes in various immunochemical markers that occur in healthy individuals of different ages. The robust age- and sex-specific reference intervals established in this study provide insight into the complex biological changes that take place throughout development and aging and will contribute to improved clinical test interpretation.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis