Merck
  • Online capillary solid-phase microextraction coupled liquid chromatography-mass spectrometry for analysis of chiral secondary alcohol products in yeast catalyzed stereoselective reduction cell culture.
Online capillary solid-phase microextraction coupled liquid chromatography-mass spectrometry for analysis of chiral secondary alcohol products in yeast catalyzed stereoselective reduction cell culture.
Journal of chromatography. A (2015-01-08)
Cheanyeh Cheng, Yu-Chuan Nian
ABSTRACT
An online solid-phase microextraction coupled liquid chromatography-electrospray ionization-ion trap mass spectrometry was developed for the analysis of trace R- and S-4-phenyl-2-butanol (R- and S-pbol) in salt rich cell culture of Saccharomyces cerevisiae catalyzed stereoselective reduction of 4-pheny-2-butanone (pbone). A Supel-Q PLOT capillary column was used for the extraction and deionized distilled water was used as the extraction mobile phase. The extraction flow rate and extraction time were at 0.1 mL min(-1) and 0.95 min, respectively. The three target analytes, pbone, R-pbol, and S-4-pbol, were desorbed and eluted by the mobile phase of water/methanol/isopropanol (55/25/20, v/v/v) with a flow rate of 0.5 mL min(-1) and analyzed by a chiral column. The mass spectrometric detection of the three target analytes was in positive ion mode with the signal [M+Na](+). The matrix-matched external standard calibration curves with linear concentration range between 0 and 50 μg mL(-1) were used for quantitative analysis. The linear regression correlation coefficients (r(2)) of the standard calibration curves were between 0.9950 and 0.9961. The yeast mediated reduction was performed with a recation culture of yeast incubation culture/glycerol (70/30, v/v) for 4 days. This biotransformation possessed 82.3% yield and 92.9% S-enantomeric excess. The limit of detection (LOD)/limit of quantification (LOQ) for pbone, R-pbol, and S-pbol was 0.02/0.067, 0.01/0.033, and 0.01/0.033 μg mL(-1), respectively. The intra-day and inter-day precisions from repeated measurements were 10.8-21.1% and 11.6-18.7%, respectively. The analysis accuracy from spike recovery was 84-91%.
Product Description
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, for use with autosampler
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol, analytical standard
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Divinylbenzene, technical grade, 55%
Sigma-Aldrich
Divinylbenzene, technical grade, 80%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), fused silica fiber, df 65 μm(PDMS/DVB, for use with manual holder, needle size 23 ga
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 24 ga
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME Portable Field Sampler, coating PDMS
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%