Merck
  • Endothelial progenitor cells and endothelial microparticles are independent predictors of endothelial function.
Endothelial progenitor cells and endothelial microparticles are independent predictors of endothelial function.
The Journal of pediatrics (2014-05-21)
Luc Bruyndonckx, Vicky Y Hoymans, Geert Frederix, Ann De Guchtenaere, Hilde Franckx, Dirk K Vissers, Christiaan J Vrints, José Ramet, Viviane M Conraads
ABSTRACT
To examine the degree of microvascular endothelial dysfunction in relation to classical cardiovascular risk factors, arterial stiffness, and numbers of circulating endothelial progenitor cells (EPCs) and endothelial microparticles (EMPs), in obese and normal-weight children. Cross-sectional study with 57 obese (15.2±1.4 years) and 30 normal-weight children (15.4±1.5 years). The principal outcome was microvascular endothelial function measured with peripheral arterial tonometry. Fasting blood samples were taken for biochemical analysis and EMPs (CD31+/CD42b- particles) and EPCs (CD34+/KDR+/CD45dim/- cells) flow cytometry. Characteristics between groups were compared by use of the appropriate independent samples test; a stepwise multiple regression analysis was used to determine independent predictors of microvascular endothelial function. Microvascular endothelial function was significantly impaired in obese children and inversely correlated with body mass index Z scores (r=-0.249; P=.021) and systolic blood pressure (r=-0.307; P=.004). The number of EPCs was significantly lower in obese children and correlated with endothelial function (r=0.250; P=.022), and the number of EMPs was significantly greater in obese children and correlated inversely with endothelial function (r=-0.255; P=.021). Multivariate analysis revealed that systolic blood pressure and numbers of circulating EPCs and EMPs are important determinants of endothelial function. Obese children demonstrate impaired endothelial microvascular function, increased arterial stiffness, fewer EPCs, and more EMPs. Besides systolic blood pressure, EPC and EMP counts independently predict the presence of microvascular endothelial dysfunction.
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Ammonium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Supelco
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Supelco
Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)