Merck
  • Analgesic effects of antidepressants alone and after their local co-administration with morphine in a rat model of neuropathic pain.
Analgesic effects of antidepressants alone and after their local co-administration with morphine in a rat model of neuropathic pain.
Pharmacological reports : PR (2014-06-07)
Grzegorz Jagla, Joanna Mika, Wioletta Makuch, Ilona Obara, Jerzy Wordliczek, Barbara Przewlocka
ABSTRACT
The therapy of neuropathic pain may include the use of co-analgesics, such as antidepressants, however, their desired analgesic effect is associated with significant side effects. An alternative approach to this is their local administration which has been proposed, but there is little data regarding their local co-administration with morphine and the nature of the interaction between morphine and either doxepin or venlafaxine, two antidepressant drugs that have been recently used in neuropathic pain therapies. This study was performed on rats after chronic constriction injury (CCI) to the sciatic nerve. The von Frey and Hargreaves' tests were used to assess mechanical allodynia and thermal hyperalgesia, respectively, after intraplantar (ipl) or subcutaneous (sc) administration of amitriptyline, doxepin, or venlafaxine, or their ipl co-administration with morphine on day 12-16 after injury. The ipl administration of amitriptyline (3, 15 mg), doxepin (1, 5, 10, 15 mg), or venlafaxine (2, 7 mg) was effective in antagonizing CCI-induced allodynia. Their sc injection at a site distal to the injured side, did not induce alterations in pain thresholds, which supports the local mode of action. Of the three antidepressants used in this study, only ipl co-administration of amitriptyline with morphine significantly enhanced its effect in contrast to doxepin and venlafaxine, both of which weakened the analgesic effect of morphine. In summary, the results suggest that when amitriptyline (but not doxepin or venlafaxine) is locally co-administered with morphine the effectiveness under neuropathic pain is enhanced, although additional studies are necessary to explain differential mechanisms of interaction of antidepressant drugs with morphine after local administration.
Sigma-Aldrich
Venlafaxine hydrochloride, ≥98% (HPLC), powder
USP
Venlafaxine hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Amitriptyline hydrochloride, ≥98% (TLC), powder
Amitriptyline hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Amitriptyline hydrochloride, United States Pharmacopeia (USP) Reference Standard
Venlafaxine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Venlafaxine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Amitriptyline hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Doxepin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Doxepin hydrochloride, ~85% E-isomer basis, ≥98% (GC), 15% Z-isomer basis, powder
Doxepin hydrochloride, European Pharmacopoeia (EP) Reference Standard