Merck
  • Catalytic function and substrate specificity of the papain-like protease domain of nsp3 from the Middle East respiratory syndrome coronavirus.
Catalytic function and substrate specificity of the papain-like protease domain of nsp3 from the Middle East respiratory syndrome coronavirus.
Journal of virology (2014-08-22)
Yahira M Báez-Santos, Anna M Mielech, Xufang Deng, Susan Baker, Andrew D Mesecar
ABSTRACT
The papain-like protease (PLpro) domain from the deadly Middle East respiratory syndrome coronavirus (MERS-CoV) was overexpressed and purified. MERS-CoV PLpro constructs with and without the putative ubiquitin-like (UBL) domain at the N terminus were found to possess protease, deubiquitinating, deISGylating, and interferon antagonism activities in transfected HEK293T cells. The quaternary structure and substrate preferences of MERS-CoV PLpro were determined and compared to those of severe acute respiratory syndrome coronavirus (SARS-CoV) PLpro, revealing prominent differences between these closely related enzymes. Steady-state kinetic analyses of purified MERS-CoV and SARS-CoV PLpros uncovered significant differences in their rates of hydrolysis of 5-aminomethyl coumarin (AMC) from C-terminally labeled peptide, ubiquitin, and ISG15 substrates, as well as in their rates of isopeptide bond cleavage of K48- and K63-linked polyubiquitin chains. MERS-CoV PLpro was found to have 8-fold and 3,500-fold higher catalytic efficiencies for hydrolysis of ISG15-AMC than for hydrolysis of the Ub-AMC and Z-RLRGG-AMC substrates, respectively. A similar trend was observed for SARS-CoV PLpro, although it was much more efficient than MERS-CoV PLpro toward ISG15-AMC and peptide-AMC substrates. MERS-CoV PLpro was found to process K48- and K63-linked polyubiquitin chains at similar rates and with similar debranching patterns, producing monoubiquitin species. However, SARS-CoV PLpro much preferred K48-linked polyubiquitin chains to K63-linked chains, and it rapidly produced di-ubiquitin molecules from K48-linked chains. Finally, potent inhibitors of SARS-CoV PLpro were found to have no effect on MERS-CoV PLpro. A homology model of the MERS-CoV PLpro structure was generated and compared to the X-ray structure of SARS-CoV PLpro to provide plausible explanations for differences in substrate and inhibitor recognition. Unlocking the secrets of how coronavirus (CoV) papain-like proteases (PLpros) perform their multifunctional roles during viral replication entails a complete mechanistic understanding of their substrate recognition and enzymatic activities. We show that the PLpro domains from the MERS and SARS coronaviruses can recognize and process the same substrates, but with different catalytic efficiencies. The differences in substrate recognition between these closely related PLpros suggest that neither enzyme can be used as a generalized model to explain the kinetic behavior of all CoV PLpros. As a consequence, decoding the mechanisms of PLpro-mediated antagonism of the host innate immune response and the development of anti-CoV PLpro enzyme inhibitors will be a challenging undertaking. The results from this study provide valuable information for understanding how MERS-CoV PLpro-mediated antagonism of the host innate immune response is orchestrated, as well as insight into the design of inhibitors against MERS-CoV PLpro.
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Z-RLRGG-AMC Acetate, ≥95% (HPLC)
SAFC
BIS-TRIS
SAFC
BIS-TRIS