Merck
  • Multiple sclerosis-associated IL2RA polymorphism controls GM-CSF production in human TH cells.
Multiple sclerosis-associated IL2RA polymorphism controls GM-CSF production in human TH cells.
Nature communications (2014-10-04)
Felix J Hartmann, Mohsen Khademi, Jehan Aram, Sandra Ammann, Ingrid Kockum, Cris Constantinescu, Bruno Gran, Fredrik Piehl, Tomas Olsson, Laura Codarri, Burkhard Becher
ABSTRACT
Genome-wide association studies implicate dysregulation of immune mechanisms in the pathogenesis of multiple sclerosis (MS). Particularly, polymorphisms in genes involved in T helper (TH) cell differentiation are associated with risk of developing MS. However, the underlying mechanism by which these risk alleles influence MS susceptibility has remained elusive. Initiation of neuroinflammation in animal models of MS has been shown to be dependent on TH cell-derived granulocyte-macrophage colony-stimulating factor (GM-CSF). We here report association of GM-CSF expression by human TH cells with MS disease severity. GM-CSF is strongly induced by interleukin 2 (IL-2). We show that an MS-associated polymorphism in the IL-2 receptor alpha (IL2RA) gene specifically increases the frequency of GM-CSF-producing TH cells. The IL2RA polymorphism regulates IL-2 responsiveness of naive TH cells and their propensity to develop into GM-CSF-producing memory TH cells. These findings mechanistically link an immunologically relevant genetic risk factor with a functional feature of TH cells in MS.
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Interleukin-2 Soluble Receptor α human, >97% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder
Sigma-Aldrich
Diethyl azodicarboxylate solution, 40 wt. % in toluene
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Pimozide
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%