Merck
  • PET imaging of disease progression and treatment effects in the experimental autoimmune encephalomyelitis rat model.
PET imaging of disease progression and treatment effects in the experimental autoimmune encephalomyelitis rat model.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2014-06-11)
Daniele de Paula Faria, Maria L H Vlaming, Sjef C V M Copray, Frans Tielen, Herma J A Anthonijsz, Jurgen W A Sijbesma, Carlos A Buchpiguel, Rudi A J O Dierckx, José W A van der Hoorn, Erik F J de Vries
ABSTRACT
The experimental autoimmune encephalomyelitis model is a model of multiple sclerosis that closely mimics the disease characteristics in humans. The main hallmarks of multiple sclerosis are neuroinflammation (microglia activation, monocyte invasion, and T-cell infiltration) and demyelination. PET imaging may be a useful noninvasive technique for monitoring disease progression and drug treatment efficacy in vivo. Experimental autoimmune encephalomyelitis was induced by myelin-oligodendrocyte glycoprotein immunization in female Dark Agouti rats. Experimental autoimmune encephalomyelitis rats were imaged at baseline and at days 6, 11, 15, and 19 after immunization to monitor monocyte and microglia activation ((11)C-PK11195) and demyelination ((11)C-MeDAS) during normal disease progression and during treatment with dexamethasone. (11)C-PK11195 PET detected activation of microglia and monocytes in the brain stem and spinal cord during disease progression. The uptake of (11)C-PK11195 was elevated in dexamethasone-treated animals that had shown mild clinical symptoms that had resolved at the time of imaging. Demyelination was not detected by (11)C-MeDAS PET, probably because of the small size of the lesions (average, 0.13 mm). PET imaging of neuroinflammation can be used to monitor disease progression and the consequences of treatment in the experimental autoimmune encephalomyelitis rat model. PET imaging was more sensitive than clinical symptoms for detecting inflammatory changes in the central nervous system.
Sigma-Aldrich
Dexamethasone 21-acetate, ≥99%
Pricing and availability is not currently available.
Dexamethasone acetate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
USP
Dexamethasone acetate, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Supelco
Dexamethasone Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.