Merck
  • Roussoella percutanea, a novel opportunistic pathogen causing subcutaneous mycoses.
Roussoella percutanea, a novel opportunistic pathogen causing subcutaneous mycoses.
Medical mycology (2014-06-28)
Sarah A Ahmed, David A Stevens, Wendy W J van de Sande, Jacques F Meis, G S de Hoog
ABSTRACT
We report the isolation of a novel agent of subcutaneous mycosis from a 45-year-old Indian male immigrant in the United States. Phylogenetic analysis of partial small ribosomal subunit and large ribosomal subunit, internal transcribed spacer, partial translation elongation factor (TEF1), and RNA polymerase second largest subunit (rPB2) loci revealed that the strain was identical to another isolate previously reported as "Madurella mycetomatis." Both strains clustered in the order Pleosporales, nested in the family Arthopyreniaceae/Roussoellaceae. The fungus differed from known species and hence a new taxon, Roussoella percutanea, is introduced, typified by a strain that showed delayed production of pycnidial conidiomata. Antifungal susceptibility testing suggested that the new species is resistant to echinocandins and flucytosine, with variable results with azoles and amphotericin B.
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Urea, SAJ first grade, ≥98.0%
Sigma-Aldrich
Urea, JIS special grade, ≥99.0%
Sigma-Aldrich
Urea, ≥99.0%
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Urea, European Pharmacopoeia (EP) Reference Standard
Supelco
Urea, analytical standard