Merck
  • Aggregation property of glycyrrhizic acid and its interaction with cyclodextrins analyzed by dynamic light scattering, isothermal titration calorimetry, and NMR.
Aggregation property of glycyrrhizic acid and its interaction with cyclodextrins analyzed by dynamic light scattering, isothermal titration calorimetry, and NMR.
Carbohydrate research (2014-05-23)
Yusuke Izutani, Kenji Kanaori, Masayuki Oda
ABSTRACT
The structural properties of glycyrrhizic acid, a sweet-tasting constituent of Glycyrrhiza glabra, and its interaction with cyclodextrins were analyzed using dynamic light scattering, isothermal titration calorimetry, and NMR. The dynamic light scattering and NMR studies showed that glycyrrhizic acid forms a water-soluble aggregate that disperses upon the addition of γ-cyclodextrin. The high sweetness of glycyrrhizic acid can be closely correlated with this aggregation, because the multimers of glycyrrhizic acid can simultaneously bind to the sweet taste receptors on the human tongue. The isothermal titration calorimetry experiments demonstrated that γ-cyclodextrin binds to glycyrrhizic acid more strongly than β-cyclodextrin, however, both reactions are accompanied by a favorable change in binding entropy. Considering the large negative change in heat capacity that is observed during the binding of γ-cyclodextrin, the main driving force for the binding is hydrophobic interactions with dehydration, which is typical for inclusion complex. NMR experiments showed that γ-cyclodextrin interacts with the central part of the aglycone moiety, not the glucuronic acid moieties, resulting in high binding affinity. It should also be noted that the two distinct complexes of glycyrrhizic acid with γ-cyclodextrin would exist in aqueous solution.
Product Description
Glycyrrhizate monoammonium, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycyrrhizic acid ammonium salt, analytical standard, suitable for HPLC
Sigma-Aldrich
Glycyrrhizic acid ammonium salt from glycyrrhiza root (licorice), ≥95.0% (NT)
Sigma-Aldrich
Glycyrrhizic acid ammonium salt from glycyrrhiza root (licorice), ≥70% (HPLC)
Ammonium glycyrrhizate, European Pharmacopoeia (EP) Reference Standard