Merck
  • Oncogenic deletion mutants of gp130 signal from intracellular compartments.
Oncogenic deletion mutants of gp130 signal from intracellular compartments.
Journal of cell science (2013-11-12)
Dirk Schmidt-Arras, Miryam Müller, Marija Stevanovic, Silke Horn, Antje Schütt, Juri Bergmann, Ruven Wilkens, Annett Lickert, Stefan Rose-John
ABSTRACT
Interleukin 6 (IL-6) and, hence, activation of the IL-6 receptor signalling subunit glycoprotein 130 (gp130; also known as interleukin-6 receptor subunit β, IL6ST), has been linked to inflammation and tumour formation. Recently, deletion mutations in gp130 have been identified in inflammatory hepatocellular adenoma. The mutations clustered around one IL-6-binding epitope and rendered gp130 constitutively active in a ligand-independent manner. Here, we show that gp130 deletion mutants, but not wild-type gp130, localise predominantly to intracellular compartments, notably the endoplasmic reticulum (ER) and early endosomes. One of the most frequent mutants, gp130 Y186-Y190del (ΔYY) is retained in the ER quality control system because of its association with the chaperone calnexin. Furthermore, we can show that gp130 ΔYY induces downstream signalling from both ER and endosomes, and that both signals contribute to ligand-independent cell proliferation. We also demonstrate that the endosomal localisation of gp130 ΔYY is crucial for fully fledged STAT3 activation. Therefore, aberrant signalling from intracellular compartments might explain the tumorigenic potential of naturally occurring somatic mutations of gp130.
Product Description
Millipore
JAK Inhibitor I, JAK Inhibitor I, CAS 457081-03-7, is a potent, reversible, cell-permeable, and ATP-competitive inhibitor of JAK 1 (IC₅₀ = 15 nM), JAK2 (IC₅₀ = 1 nM), JAK3 (Ki = 5 nM) and Tyk2 (IC₅₀ = 1 nM).