Merck
  • Highly modular bow-tie gene circuits with programmable dynamic behaviour.
Highly modular bow-tie gene circuits with programmable dynamic behaviour.
Nature communications (2014-10-15)
Laura Prochazka, Bartolomeo Angelici, Benjamin Haefliger, Yaakov Benenson
ABSTRACT
Synthetic gene circuits often require extensive mutual optimization of their components for successful operation, while modular and programmable design platforms are rare. A possible solution lies in the 'bow-tie' architecture, which stipulates a focal component-a 'knot'-uncoupling circuits' inputs and outputs, simplifying component swapping, and introducing additional layer of control. Here we construct, in cultured human cells, synthetic bow-tie circuits that transduce microRNA inputs into protein outputs with independently programmable logical and dynamic behaviour. The latter is adjusted via two different knot configurations: a transcriptional activator causing the outputs to track input changes reversibly, and a recombinase-based cascade, converting transient inputs into permanent actuation. We characterize the circuits in HEK293 cells, confirming their modularity and scalability, and validate them using endogenous microRNA inputs in additional cell lines. This platform can be used for biotechnological and biomedical applications in vitro, in vivo and potentially in human therapy.
Product Description
Supelco
Doxycycline hyclate, VETRANAL®, analytical standard
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Supelco
Doxycycline hyclate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Doxycycline hyclate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Doxycycline hyclate, European Pharmacopoeia (EP) Reference Standard
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
Streptomycin Ready Made Solution, 100 mg/mL in water