Merck
  • A phase 3 non-inferiority study of 5-FU/l-leucovorin/irinotecan (FOLFIRI) versus irinotecan/S-1 (IRIS) as second-line chemotherapy for metastatic colorectal cancer: updated results of the FIRIS study.
A phase 3 non-inferiority study of 5-FU/l-leucovorin/irinotecan (FOLFIRI) versus irinotecan/S-1 (IRIS) as second-line chemotherapy for metastatic colorectal cancer: updated results of the FIRIS study.
Journal of cancer research and clinical oncology (2014-08-12)
Hirofumi Yasui, Kei Muro, Yasuhiro Shimada, Akihito Tsuji, Shinichi Sameshima, Hideo Baba, Taroh Satoh, Tadamichi Denda, Kenji Ina, Tomohiro Nishina, Kensei Yamaguchi, Taito Esaki, Shinya Tokunaga, Hiroyuki Kuwano, Narikazu Boku, Yoshito Komatsu, Masahiko Watanabe, Ichinosuke Hyodo, Satoshi Morita, Kenichi Sugihara
ABSTRACT
The FIRIS study previously demonstrated non-inferiority of IRIS (irinotecan plus S-1) to FOLFIRI (5-fluorouracil/leucovorin with irinotecan) for progression-free survival as the second-line chemotherapy for metastatic colorectal cancer (mCRC) as the primary endpoint. The overall survival (OS) data were immature at the time of the primary analysis. Between 30 January 2006 and 29 January 2008, 426 patients with mCRC who failed in first-line chemotherapy were randomly assigned to receive either FOLFIRI or IRIS. After the primary analysis, the follow-up survey was cut off on 29 July 2010, and the final OS data were analysed. With a median follow-up of 39.2 months, the median OS was 17.4 months in the FOLFIRI group and 17.8 months in the IRIS group [hazard ratio (HR) 0.900; 95% confidence interval (CI) 0.728-1.112]. In the pre-planned subgroup of patients who received prior chemotherapy containing oxaliplatin, the median OS was 12.7 months in the FOLFIRI group and 15.3 months in the IRIS group (HR 0.755; 95% CI 0.580-0.983). IRIS is non-inferior to FOLFIRI for OS as second-line chemotherapy for mCRC. IRIS can be an option for second-line chemotherapy of mCRC. (ClinicalTrials.gov Number: NCT00284258).
Sigma-Aldrich
7-Ethyl-10-hydroxycamptothecin, ≥98% (HPLC), powder
Sigma-Aldrich
Oxaliplatin, powder
Oxaliplatin, European Pharmacopoeia (EP) Reference Standard
Calcium folinate, European Pharmacopoeia (EP) Reference Standard
Supelco
Oxaliplatin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Oxaliplatin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Folinic acid calcium salt hydrate, BioXtra, ≥99.0% (HPLC)
Sigma-Aldrich
Irinotecan hydrochloride, topoisomerase inhibitor
Supelco
Folinic acid calcium salt hydrate, analytical standard
Sigma-Aldrich
Tegafur, ≥98% (HPLC), powder