Merck
Ki67 and proliferation in breast cancer.
Journal of clinical pathology (2013-02-26)
Nirmala Pathmanathan, Rosemary L Balleine
ABSTRACT
New approaches to the prognostic assessment of breast cancer have come from molecular profiling studies. A major feature of this work has been to emphasise the importance of cancer cell proliferation as a key discriminative indicator of recurrence risk for oestrogen receptor positive breast cancer in particular. Mitotic count scoring, as a component of histopathological grade, has long formed part of a routine evaluation of breast cancer biology. However, there is an increasingly compelling case to include a specific proliferation score in breast cancer pathology reports based on expression of the cell cycle regulated protein Ki67. Immunohistochemical staining for Ki67 is a widely available and economical test with good tolerance of pre-analytical variations and staining conditions. However, there is currently no evidence based protocol established to derive a reliable and informative Ki67 score for routine clinical use. In this circumstance, pathologists must establish a standardised framework for scoring Ki67 and communicating results to a multidisciplinary team.
Sigma-Aldrich
Ki-67 (SP6) Rabbit Monoclonal Antibody
Pricing and availability is not currently available.