Merck
  • Novel N-phenyl-substituted thiazolidinediones protect neural cells against glutamate- and tBid-induced toxicity.
Novel N-phenyl-substituted thiazolidinediones protect neural cells against glutamate- and tBid-induced toxicity.
The Journal of pharmacology and experimental therapeutics (2014-05-23)
Sina Oppermann, Florian C Schrader, Katharina Elsässer, Amalia M Dolga, Anna Lena Kraus, Nunzianna Doti, Christof Wegscheid-Gerlach, Martin Schlitzer, Carsten Culmsee
ABSTRACT
Mitochondrial demise is a key feature of progressive neuronal death contributing to acute and chronic neurological disorders. Recent studies identified a pivotal role for the BH3-only protein B-cell lymphoma-2 interacting domain death antagonist (Bid) for such mitochondrial damage and delayed neuronal death after oxygen-glucose deprivation, glutamate-induced excitotoxicity, or oxidative stress in vitro and after cerebral ischemia in vivo. Therefore, we developed new N-phenyl-substituted thiazolidine-2,4-dione derivatives as potent inhibitors of Bid-dependent neurotoxicity. The new compounds 6, 7, and 16 were identified as highly protective by extensive screening in a model of glutamate toxicity in immortalized mouse hippocampal neurons (HT-22 cells). These compounds significantly prevent truncated Bid-induced toxicity in the neuronal cell line, providing strong evidence that inhibition of Bid was the underlying mechanism of the observed protective effects. Furthermore, Bid-dependent hallmarks of mitochondrial dysfunction, such as loss of mitochondrial membrane potential, ATP depletion, as well as impairments in mitochondrial respiration, are significantly prevented by compounds 6, 7, and 16. Therefore, the present study identifies a class of N-phenyl thiazolidinediones as novel Bid-inhibiting neuroprotective agents that provide promising therapeutic perspectives for neurodegenerative diseases, in which Bid-mediated mitochondrial damage and associated intrinsic death pathways contribute to the underlying progressive loss of neurons.
Sigma-Aldrich
Butyramide, ≥98.0% (T)
Sigma-Aldrich
L-Glutamic acid hydrochloride
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamic acid, JIS special grade, ≥99.0%
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
D-Glutamic acid, ≥99% (TLC)
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
BI-6C9, ≥97% (HPLC), solid
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland