Merck
Molybdenum metabolism in plants.
Metallomics : integrated biometal science (2013-06-27)
Manuel Tejada-Jiménez, Alejandro Chamizo-Ampudia, Aurora Galván, Emilio Fernández, Ángel Llamas
ABSTRACT
The viability of plants relies on molybdenum, which after binding to the organic moiety of molybdopterin forms the molybdenum cofactor (Moco) and acquires remarkable redox properties. Moco is in the active site of critical molybdoenzymes, which use to work as small electron transport chains and participate in N and S metabolism, hormone biosynthesis, toxic compound transformations and other important processes not only in plants but also in all the other kingdoms of life. Molybdate metabolism in plants is reviewed here, with special attention to two main aspects, the different molybdate transporters that with a very high affinity participate in molybdenum acquisition and the recently discovered Moco enzyme amidoxime-reducing component. Their functionality is starting to be understood.
Sigma-Aldrich
Molybdenum, powder, 1-5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Molybdenum, powder, 10 μm, ≥99.95% trace metals basis
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Molybdenum, powder, <150 μm, 99.9% trace metals basis
Sigma-Aldrich
Molybdenum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Molybdenum, foil, light tested, 25x25mm, thickness 0.05mm, annealed, 99.9%
Molybdenum, foil, not light tested, 25x25mm, thickness 0.005mm, 99.9%
Molybdenum, foil, light tested, 50x50mm, thickness 0.009mm, 99.9%
Molybdenum, foil, 1m coil, thickness 0.008mm, 99.9%
Molybdenum, foil, 50mm disks, thickness 0.30mm, annealed, 99.9%
Molybdenum, foil, 15mm disks, thickness 0.025mm, annealed, 99.9%
Molybdenum, foil, 4mm disks, thickness 0.15mm, annealed, 99.9%
Molybdenum, foil, 10mm disks, thickness 0.05mm, annealed, 99.9%
Molybdenum, foil, 6mm disks, thickness 0.03mm, annealed, 99.9%
Molybdenum, rod, 100mm, diameter 25mm, centerless ground, 99.9%
Molybdenum, rod, 200mm, diameter 5.0mm, centerless ground, 99.9%
Molybdenum, foil, 50mm disks, thickness 0.01mm, 99.9%
Molybdenum, foil, 4mm disks, thickness 0.01mm, 99.9%
Molybdenum, foil, 25mm disks, thickness 0.0125mm, 99.9%
Molybdenum, foil, 50mm disks, thickness 0.015mm, as rolled, 99.9%
Molybdenum, foil, 6mm disks, thickness 0.075mm, annealed, 99.9%
Molybdenum, foil, 25mm disks, thickness 0.05mm, annealed, 99.9%
Molybdenum, foil, 50mm disks, thickness 0.25mm, annealed, 99.9%
Molybdenum, foil, 4mm disks, thickness 0.005mm, 99.9%
Molybdenum, foil, 25mm disks, thickness 0.015mm, annealed, 99.9%
Molybdenum, foil, 6mm disks, thickness 0.020mm, annealed, 99.9%
Molybdenum, foil, 50mm disks, thickness 0.125mm, annealed, 99.9%